Assay Details
| Gene Symbol: STPG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001918067.4 | XP_001918102.2 | 318 | 72 | ||
| XM_005605965.3 | XP_005606022.2 | 318 | 72 | |||
| XM_005605966.3 | XP_005606023.2 | 318 | 72 | |||
More Information
Set Membership: |
|

STPG3
Genomic Map