Assay Details
| Gene Symbol: BACH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023629987.1 | XP_023485755.1 | 417 | 83 | ||
| XM_023629988.1 | XP_023485756.1 | 468 | 83 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BACH1| Gene Symbol: BACH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023629987.1 | XP_023485755.1 | 417 | 83 | ||
| XM_023629988.1 | XP_023485756.1 | 468 | 83 | |||
Set Membership: |
|