Assay Details
| Gene Symbol: MRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736334.2 | XP_014591820.1 | 299 | 77 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736334.2 | XP_014591820.1 | 299 | 77 | ||