Assay Details
| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492982.5 | XP_001493032.3 | 293 | 60 | ||
| XM_023630141.1 | XP_023485909.1 | 293 | 60 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VPS26C| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492982.5 | XP_001493032.3 | 293 | 60 | ||
| XM_023630141.1 | XP_023485909.1 | 293 | 60 | |||
Set Membership: |
|