Assay Details
| Gene Symbol: KCNJ15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736377.2 | XP_014591863.1 | 2778 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014736377.2 transcripts.
Set Membership: |
|

KCNJ15
Genomic Map