Assay Details
| Gene Symbol: PCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606157.3 | XP_005606214.1 | 128 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005606157.3 transcripts.
Set Membership: |
|

PCP4
Genomic Map