Assay Details
Gene Symbol: CFAP410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490381.4 | XP_001490431.2 | 232 | 60 | ||
XM_014736418.2 | XP_014591904.2 | 234 | 60 | |||
XM_023630275.1 | XP_023486043.1 | 216 | 60 | |||
XM_023630276.1 | XP_023486044.1 | 214 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
