Assay Details
| Gene Symbol: MFHAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023630576.1 | XP_023486344.1 | 3418 | 71 | ||
| XM_023630580.1 | XP_023486348.1 | 3263 | 71 | |||
More Information
Set Membership: |
|

MFHAS1
Genomic Map