Assay Details
| Gene Symbol: SNRPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003364272.4 | XP_003364320.1 | 159 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003364272.4 transcripts.
Set Membership: |
|

SNRPF
Genomic Map