Assay Details
| Gene Symbol: WASHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497456.4 | XP_001497506.1 | 163 | 80 | ||
| XM_005606547.2 | XP_005606604.1 | 163 | 80 | |||
| XM_023631061.1 | XP_023486829.1 | 163 | 80 | |||
More Information
Set Membership: |
|

WASHC3
Genomic Map