Assay Details
| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736782.2 | XP_014592268.1 | 2850 | 73 | ||
| XM_023631814.1 | XP_023487582.1 | 2850 | 73 | |||
| XM_023631815.1 | XP_023487583.1 | 2742 | 73 | |||
| XM_023631816.1 | XP_023487584.1 | 2715 | 73 | |||
| XM_023631817.1 | XP_023487585.1 | 2329 | 73 | |||

WASHC4
Genomic Map