Assay Details
| Gene Symbol: SSTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606611.3 | XP_005606668.2 | 10612 | 88 | ||
| XM_023631863.1 | XP_023487631.1 | 10613 | 88 | |||
| XM_023631864.1 | XP_023487632.1 | 10613 | 88 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005606611.3, XM_023631863.1, XM_023631864.1 transcripts.
Set Membership: |
|

SSTR3
Genomic Map