Assay Details
| Gene Symbol: C28H22orf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001916464.4 | XP_001916499.3 | 170 | 106 | ||
| XM_023631330.1 | XP_023487098.1 | 172 | 106 | |||
More Information
Set Membership: |
|

C28H22orf23
Genomic Map