Assay Details
Gene Symbol: LOC100055155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606631.3 | XP_005606688.1 | 201 | 69 | ||
XM_014736832.2 | XP_014592318.1 | 406 | 69 | |||
XM_014736833.2 | XP_014592319.1 | 103 | 69 | |||
XM_014736834.2 | XP_014592320.1 | 866 | 69 | |||
XM_023631663.1 | XP_023487431.1 | 221 | 69 | |||
XM_023631666.1 | XP_023487434.1 | 221 | 69 | |||
XM_023631668.1 | XP_023487436.1 | 221 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
