Assay Details
| Gene Symbol: LOC100055155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606631.3 | XP_005606688.1 | 1270 | 63 | ||
| XM_014736832.2 | XP_014592318.1 | 1475 | 63 | |||
| XM_014736833.2 | XP_014592319.1 | 1172 | 63 | |||
| XM_014736834.2 | XP_014592320.1 | 1935 | 63 | |||
| XM_023631663.1 | XP_023487431.1 | 1290 | 63 | |||
| XM_023631666.1 | XP_023487434.1 | 1229 | 63 | |||
| XM_023631667.1 | XP_023487435.1 | 1277 | 63 | |||
More Information
Set Membership: |
|

LOC100055155
Genomic Map