Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606696.3 | XP_005606753.3 | 2445 | 95 | ||
| XM_014736869.2 | XP_014592355.2 | 2445 | 95 | |||
| XM_023631047.1 | XP_023486815.1 | 2443 | 95 | |||
| XM_023631048.1 | XP_023486816.1 | 2445 | 95 | |||
| XM_023631049.1 | XP_023486817.1 | 1096 | 95 | |||
| XM_023631050.1 | XP_023486818.1 | 1178 | 95 | |||
More Information
Set Membership: |
|

SEPTIN3
Genomic Map