Assay Details
| Gene Symbol: BIK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736880.2 | XP_014592366.1 | 387 | 81 | ||
| XM_023631183.1 | XP_023486951.1 | 3587 | 81 | |||
| XM_023631184.1 | XP_023486952.1 | 3583 | 81 | |||
| XM_023631185.1 | XP_023486953.1 | 1565 | 81 | |||
More Information
Set Membership: |
|

BIK
Genomic Map