Assay Details
| Gene Symbol: KLF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606962.3 | XP_005607019.1 | 1043 | 125 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KLF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005606962.3 | XP_005607019.1 | 1043 | 125 | ||
Set Membership: |
|