Assay Details
| Gene Symbol: TENT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023636936.1 | XP_023492704.1 | 915 | 116 | ||
| XM_023636937.1 | XP_023492705.1 | 915 | 116 | |||
More Information
Set Membership: |
|

TENT4B
Genomic Map