Assay Details
| Gene Symbol: RIPOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638631.1 | XP_023494399.1 | 3915 | 80 | ||
| XM_023638632.1 | XP_023494400.1 | 3888 | 80 | |||
| XM_023638633.1 | XP_023494401.1 | 3881 | 80 | |||
| XM_023638634.1 | XP_023494402.1 | 4126 | 80 | |||
| XM_023638635.1 | XP_023494403.1 | 3837 | 80 | |||
| XM_023638636.1 | XP_023494404.1 | 3651 | 80 | |||
| XM_023638637.1 | XP_023494405.1 | 3816 | 80 | |||
| XM_023638638.1 | XP_023494406.1 | 3912 | 80 | |||
| XM_023638639.1 | XP_023494407.1 | 4426 | 80 | |||
| XM_023638640.1 | XP_023494408.1 | 3188 | 80 | |||
More Information
Set Membership: |
|

RIPOR1
Genomic Map