Assay Details
| Gene Symbol: LOC100068023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497923.5 | XP_001497973.2 | 1008 | 92 | ||
| XM_005608398.3 | XP_005608455.1 | 1049 | 92 | |||
| XM_023637263.1 | XP_023493031.1 | 717 | 92 | |||
| XM_023637264.1 | XP_023493032.1 | 988 | 92 | |||
| XM_023637265.1 | XP_023493033.1 | 812 | 92 | |||
| XM_023637266.1 | XP_023493034.1 | 845 | 92 | |||
| XM_023637267.1 | XP_023493035.1 | 578 | 92 | |||

LOC100068023
Genomic Map