Assay Details
| Gene Symbol: LOC100068023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497923.5 | XP_001497973.2 | 1387 | 78 | ||
| XM_005608398.3 | XP_005608455.1 | 1428 | 78 | |||
| XM_005608399.3 | XP_005608456.1 | 1344 | 78 | |||
| XM_023637263.1 | XP_023493031.1 | 1096 | 78 | |||
| XM_023637264.1 | XP_023493032.1 | 1367 | 78 | |||
| XM_023637265.1 | XP_023493033.1 | 1191 | 78 | |||
| XM_023637266.1 | XP_023493034.1 | 1224 | 78 | |||
| XM_023637267.1 | XP_023493035.1 | 957 | 78 | |||
| GenBank mRNA | JL625812.1 | - | 3 - 4 | 370 | 78 | |
More Information
Set Membership: |
|

LOC100068023
Genomic Map