Assay Details
| Gene Symbol: LOC100054938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500964.5 | XP_001501014.1 | 198 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100054938| Gene Symbol: LOC100054938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500964.5 | XP_001501014.1 | 198 | 73 | ||