Assay Details
| Gene Symbol: CIBAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023637433.1 | XP_023493201.1 | 560 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CIBAR2| Gene Symbol: CIBAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023637433.1 | XP_023493201.1 | 560 | 84 | ||