Assay Details
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494130.3 | XP_001494180.1 | 401 | 69 | ||
| GenBank mRNA | JL616002.1 | - | 2 - 3 | 326 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494130.3 | XP_001494180.1 | 401 | 69 | ||
| GenBank mRNA | JL616002.1 | - | 2 - 3 | 326 | 69 | |
Set Membership: |
|