Assay Details
| Gene Symbol: PWWP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014741248.2 | XP_014596734.1 | 1064 | 84 | ||
| XM_023646292.1 | XP_023502060.1 | 1139 | 84 | |||
| XM_023646293.1 | XP_023502061.1 | 1237 | 84 | |||
| XM_023646294.1 | XP_023502062.1 | 1316 | 84 | |||
| XM_023646295.1 | XP_023502063.1 | 1391 | 84 | |||
| XM_023646296.1 | XP_023502064.1 | 1138 | 84 | |||
| XM_023646297.1 | XP_023502065.1 | 1639 | 84 | |||
| XM_023646298.1 | XP_023502066.1 | 1768 | 84 | |||
| XM_023646299.1 | XP_023502067.1 | 1004 | 84 | |||
| XM_023646300.1 | XP_023502068.1 | 1015 | 84 | |||
More Information
Set Membership: |
|

PWWP3A
Genomic Map