Assay Details
| Gene Symbol: PWWP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014741248.2 | XP_014596734.1 | 1242 | 91 | ||
| XM_023646292.1 | XP_023502060.1 | 1317 | 91 | |||
| XM_023646293.1 | XP_023502061.1 | 1415 | 91 | |||
| XM_023646294.1 | XP_023502062.1 | 1494 | 91 | |||
| XM_023646295.1 | XP_023502063.1 | 1569 | 91 | |||
| XM_023646296.1 | XP_023502064.1 | 1316 | 91 | |||
| XM_023646297.1 | XP_023502065.1 | 1817 | 91 | |||
| XM_023646298.1 | XP_023502066.1 | 1946 | 91 | |||
| XM_023646299.1 | XP_023502067.1 | 1182 | 91 | |||
| XM_023646300.1 | XP_023502068.1 | 1193 | 91 | |||
More Information
Set Membership: |
|

PWWP3A
Genomic Map