Assay Details
| Gene Symbol: PLAC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023637811.1 | XP_023493579.1 | 511 | 105 | ||
| GenBank mRNA | JL628065.1 | - | 1 - 2 | 187 | 105 | |
More Information
Set Membership: |
|

PLAC8A
Genomic Map