Assay Details
Gene Symbol: LOC100630489 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364674.4 | XP_003364722.1 | 367 | 76 | ||
XM_005608718.3 | XP_005608775.1 | 358 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
