Assay Details
Gene Symbol: NSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014738599.2 | XP_014594085.1 | 1467 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014738599.2 transcripts.
Set Membership: |
![]() ![]() |
