Assay Details
Gene Symbol: NSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638511.1 | XP_023494279.1 | 1711 | 72 | ||
XM_023638512.1 | XP_023494280.1 | 1711 | 72 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023638511.1 | XP_023494279.1 | 1711 | 72 | ||
XM_023638512.1 | XP_023494280.1 | 1711 | 72 |