Assay Details
| Gene Symbol: LOC100050174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738055.2 | XP_014593541.1 | 753 | 90 | ||
| XM_014738056.2 | XP_014593542.1 | 740 | 90 | |||
| XM_014738058.2 | XP_014593544.1 | 580 | 90 | |||
| XM_023632759.1 | XP_023488527.1 | 788 | 90 | |||
More Information
Set Membership: |
|

LOC100050174
Genomic Map