Assay Details
| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488930.6 | XP_001488980.1 | 3668 | 106 | ||
| XM_023632724.1 | XP_023488492.1 | 3690 | 106 | |||
| XM_023632725.1 | XP_023488493.1 | 3691 | 106 | |||
More Information
Set Membership: |
|

EPRS1
Genomic Map