Assay Details
| Gene Symbol: CCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489474.6 | XP_001489524.3 | 243 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001489474.6 transcripts.
Set Membership: |
|

CCR6
Genomic Map