Assay Details
| Gene Symbol: UBE2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638978.1 | XP_023494746.1 | 330 | 98 | ||
| XM_023638980.1 | XP_023494748.1 | 330 | 98 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBE2D4| Gene Symbol: UBE2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638978.1 | XP_023494746.1 | 330 | 98 | ||
| XM_023638980.1 | XP_023494748.1 | 330 | 98 | |||
Set Membership: |
|