Assay Details
| Gene Symbol: MAML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497899.6 | XP_001497949.3 | 3376 | 90 | ||
| XM_023646427.1 | XP_023502195.1 | 11537 | 90 | |||
More Information
Set Membership: |
|

MAML2
Genomic Map