Assay Details
| Gene Symbol: LOC100069290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014741350.2 | XP_014596836.2 | 588 | 106 | ||
| XM_023644724.1 | XP_023500492.1 | 637 | 106 | |||
More Information
Set Membership: |
|

LOC100069290
Genomic Map