Assay Details
| Gene Symbol: POGLUT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023644750.1 | XP_023500518.1 | 1148 | 92 | ||
| GenBank mRNA | JL639837.1 | - | 1 - 2 | 112 | 92 | |
More Information
Set Membership: |
|

POGLUT3
Genomic Map