Assay Details
| Gene Symbol: ADGRE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645194.1 | XP_023500962.1 | 319 | 184 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADGRE3| Gene Symbol: ADGRE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645194.1 | XP_023500962.1 | 319 | 184 | ||
Set Membership: |
|