Assay Details
| Gene Symbol: ADGRE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005611833.3 | XP_005611890.1 | 592 | 100 | ||
| XM_023645192.1 | XP_023500960.1 | 607 | 100 | |||
| XM_023645193.1 | XP_023500961.1 | 823 | 100 | |||
| XM_023645194.1 | XP_023500962.1 | 450 | 100 | |||
More Information
Set Membership: |
|

ADGRE3
Genomic Map