Assay Details
| Gene Symbol: ODAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645361.1 | XP_023501129.1 | 381 | 69 | ||
| XM_023645362.1 | XP_023501130.1 | 382 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ODAD3| Gene Symbol: ODAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645361.1 | XP_023501129.1 | 381 | 69 | ||
| XM_023645362.1 | XP_023501130.1 | 382 | 69 | |||
Set Membership: |
|