Assay Details
| Gene Symbol: LOC100061960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645504.1 | XP_023501272.1 | 86 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023645504.1 transcripts.
Set Membership: |
|

LOC100061960
Genomic Map