Assay Details
| Gene Symbol: LOC100061960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014741873.2 | XP_014597359.2 | 485 | 85 | ||
| XM_023645504.1 | XP_023501272.1 | 374 | 85 | |||
More Information
Set Membership: |
|

LOC100061960
Genomic Map