Assay Details
| Gene Symbol: NDUFA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023639283.1 | XP_023495051.1 | 161 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023639283.1 transcripts.
Set Membership: |
|

NDUFA4
Genomic Map