Assay Details
Gene Symbol: CRPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639316.1 | XP_023495084.1 | 992 | 101 | ||
XM_023639317.1 | XP_023495085.1 | 992 | 101 | |||
XM_023639319.1 | XP_023495087.1 | 992 | 101 | |||
XM_023639320.1 | XP_023495088.1 | 992 | 101 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
