Assay Details
| Gene Symbol: BPGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500706.6 | XP_001500756.1 | 129 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001500706.6 transcripts.
Set Membership: |
|

BPGM
Genomic Map