Assay Details
| Gene Symbol: IRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501061.6 | XP_001501111.4 | 1096 | 67 | ||
| XM_014741853.2 | XP_014597339.1 | 1092 | 67 | |||
| XM_014741854.2 | XP_014597340.1 | 784 | 67 | |||
More Information
Set Membership: |
|

IRAG1
Genomic Map