Assay Details
Gene Symbol: LOC100072189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646186.1 | XP_023501954.1 | 785 | 102 | ||
XM_023646187.1 | XP_023501955.1 | 789 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
