Assay Details
| Gene Symbol: WT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646212.1 | XP_023501980.1 | 1227 | 71 | ||
| XM_023646214.1 | XP_023501982.1 | 980 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: WT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646212.1 | XP_023501980.1 | 1227 | 71 | ||
| XM_023646214.1 | XP_023501982.1 | 980 | 71 | |||
Set Membership: |
|