Assay Details
Gene Symbol: PISD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494448.6 | XP_001494498.3 | 550 | 82 | ||
XM_005612418.3 | XP_005612475.1 | 6307 | 82 | |||
XM_014742030.2 | XP_014597516.1 | 490 | 82 | |||
XM_014742031.2 | XP_014597517.1 | 619 | 82 | |||
XM_023646833.1 | XP_023502601.1 | 469 | 82 | |||
XM_023646835.1 | XP_023502603.1 | 347 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
